missing translation for 'onlineSavingsMsg'
Learn More

ZNF394 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25785-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16874345

  • 4593.76 NOK / 150µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
Specifications

Specifications

ZNF394
Polyclonal
Unconjugated
ZNF394
zinc finger protein 394, ZKSCAN14, ZNF394
Rabbit
Antigen Affinity Chromatography
RUO
67235, 84124
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q53GI3, Q9Z1D9
Znf394
ZNF394 Fusion Protein Ag22655
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers