missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF382 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26560-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the C2H2 zinc finger transcription factor family, such as ZNF382, play key roles in the regulation of cell proliferation, differentiation, and apoptosis in response to a variety of stimuli. Members of the C2H2 zinc finger transcription factor family, such as ZNF382, play key roles in the regulation of cell proliferation, differentiation, and apoptosis in response to a variety of stimuli (Gebelein et al., 1998).Specifications
| ZNF382 | |
| Polyclonal | |
| Unconjugated | |
| ZNF382 | |
| KS1, zinc finger protein 382, ZNF382 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 233060, 84911 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B2RXC5, Q96SR6 | |
| ZNF382 | |
| ZNF382 Fusion Protein Ag23348 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction