missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF232 Antibody (PCRP-ZNF232-1D5) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24040JF635
This item is not returnable.
View return policy
Description
ZNF232 Monoclonal antibody specifically detects ZNF232 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)
Specifications
| ZNF232 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| zinc finger protein 500, Zinc finger protein with KRAB and SCAN domains 18, ZKSCAN18KIAA0557 | |
| Recombinant fragment (around aa29-109) of human ZNF232 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-ZNF232-1D5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7775 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction