Learn More
ZNF215 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25502-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16884065
  • 1611.84 NOK / 20µL
Estimated Shipment: 23-05-2024
Description
May be involved in transcriptional regulation.
Specifications
Specifications
ZNF215
Polyclonal
Unconjugated
ZNF215
BAZ 2, BAZ2, zinc finger protein 215, ZKSCAN11, ZNF215
Rabbit
Antigen Affinity Chromatography
RUO
7762
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UL58
ZNF215
ZNF215 Fusion Protein Ag22026
20 μL
Primary
Mouse, Human