missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF215 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25502-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May be involved in transcriptional regulation.Specifications
ZNF215 | |
Polyclonal | |
Unconjugated | |
ZNF215 | |
BAZ 2, BAZ2, zinc finger protein 215, ZKSCAN11, ZNF215 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7762 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UL58 | |
ZNF215 | |
ZNF215 Fusion Protein Ag22026 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |