Learn More
ZMYND12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25587-1-AP-20UL
 View more versions of this product
Product Code. 16844145
  • 1710.33 NOK / 20µL
Description
ZMYND12 is a protein coding gene.
Specifications
Specifications
ZMYND12
Polyclonal
Unconjugated
ZMYND12
ZMYND12
Rabbit
Antigen Affinity Chromatography
RUO
313552, 332934, 84217
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H0C1
Zmynd12
ZMYND12 Fusion Protein Ag22358
20 μL
Primary
Rat, Human, Mouse