Learn More
ZMAT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10504-1-AP-150UL
 View more versions of this product
Product Code. 16886923
  • 4836.10 NOK / 150µL
Description
This gene encodes a protein containing three zinc finger domains and a nuclear localization signal. The mRNA and the protein of this gene are upregulated by wildtype p53 and overexpression of this gene inhibits tumor cell growth, suggesting that this gene may have a role in the p53-dependent growth regulatory pathway. Alternative splicing of this gene results in two transcript variants encoding two isoforms differing in only one amino acid.
Specifications
Specifications
ZMAT3
Polyclonal
Unconjugated
ZMAT3
p53 activated gene 608 protein, PAG608, WIG 1, WIG1, Zinc finger protein WIG 1, zinc finger, matrin type 3, ZMAT3
Rabbit
Antigen Affinity Chromatography
RUO
22401, 64393, 64394
-20°C
Liquid
Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08781, O54836, Q9HA38
ZMAT3
ZMAT3 Fusion Protein Ag0780
150 μL
Primary
Human, Mouse, Rat