Learn More
Thermo Scientific Chemicals Zirconium Oxide Round Bottom Crucibles, High Form
Click to view available options
Capacity (Metric):
5 mL
10 mL
100 mL
200 mL
Diameter (Metric) Outer:
26mm
30mm
56mm
69mm
Height (Metric):
29mm
34mm
68mm
85mm
Förpackningsstorlek
Each
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Material | Zirconium Oxide |
| Quantity | 1 Piece |
| Capacity (Metric) | 5 mL |
| Diameter (Metric) Outer | 26mm |
| Height (Metric) | 29mm |
| Product Type | Zirconium Oxide Round Bottom Crucibles, High Form |
| Diameter (Metric) Base | 18mm |
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion