Learn More
Zirconium(IV) Acetylacetonate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 17501-44-9 |
| Melting Point | 189°C to 192°C |
| MDL Number | MFCD00000036 |
| Quantity | 100 g |
| Synonym | Zirconium(IV) acetylacetonate, Tetrakis(2,4-pentanedionato)zirconium(IV), Tetrakis(acetylacetonato)zirconium(IV), Zirconium(IV) acetylacetonate |
| Molecular Weight (g/mol) | 487.66 |
| Grade | Research |