Learn More
Thermo Scientific Chemicals Zirconium Cover for Crucible
Product Code. 15431407
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
15 mL
20 mL
25 mL
35 mL
45 mL
55 mL
100 mL
250 mL
500 mL
1000 mL
Description:
For 35884 crucible
For 35888 crucible
For 35905 crucible
For 35909 crucible
For 35926 crucible
For 35930 crucible
For 35947 crucible
For 35951 crucible
For 35968 crucible
For 35972 crucible
For 35997 crucible
For 36014 crucible
For 36035 crucible
For 36039 crucible
For 36054 crucible
For 36058 crucible
For 36070 crucible
For 36083 crucible
For 36087 crucible
For 36097 crucible
Unit Size:
Each
Product Code. Capacity (Metric) Description unitSize
15431407 45 mL For 35968 crucible Each
15471357 1000 mL For 36035 crucible Each
15401367 500 mL For 36039 crucible Each
15451367 500 mL For 36087 crucible Each
15481367 250 mL For 36058 crucible Each
15431377 250 mL For 36097 crucible Each
15461377 100 mL For 36083 crucible Each
15411387 100 mL For 36054 crucible Each
15421397 55 mL For 35997 crucible Each
15451397 15 mL For 35884 crucible Each
15481397 5 mL For 35888 crucible Each
15461407 55 mL For 36070 crucible Each
15401417 20 mL For 35905 crucible Each
15431417 10 mL For 35909 crucible Each
15471417 35 mL For 35947 crucible Each
15401427 25 mL For 35972 crucible Each
15441427 25 mL For 35926 crucible Each
15481427 15 mL For 35930 crucible Each
15481437 20 mL For 35951 crucible Each
15401447 45 mL For 36014 crucible Each
20 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15431407
Brand: Thermo Scientific Chemicals 036022.KT
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description For 35968 crucible
Material Zirconium
Capacity (Metric) 45 mL
Height (Metric) Depth:22mm
Product Type Zirconium Crucible Cover