Learn More
- Home
- Products
- Specialty Lab Equipment, Instruments, and Apparatuses
- Crucibles
- Thermo Scientific Chemicals Zirconium Cover for Crucible
Thermo Scientific Chemicals Zirconium Cover for Crucible
Product Code. 15401447| Product Code. | Capacity (Metric) | Description | unitSize |
|---|---|---|---|
| 15401447 | 45 mL | For 36014 crucible | Each |
| 15471357 | 1000 mL | For 36035 crucible | Each |
| 15401367 | 500 mL | For 36039 crucible | Each |
| 15451367 | 500 mL | For 36087 crucible | Each |
| 15481367 | 250 mL | For 36058 crucible | Each |
| 15431377 | 250 mL | For 36097 crucible | Each |
| 15461377 | 100 mL | For 36083 crucible | Each |
| 15411387 | 100 mL | For 36054 crucible | Each |
| 15421397 | 55 mL | For 35997 crucible | Each |
| 15451397 | 15 mL | For 35884 crucible | Each |
| 15481397 | 5 mL | For 35888 crucible | Each |
| 15431407 | 45 mL | For 35968 crucible | Each |
| 15461407 | 55 mL | For 36070 crucible | Each |
| 15401417 | 20 mL | For 35905 crucible | Each |
| 15431417 | 10 mL | For 35909 crucible | Each |
| 15471417 | 35 mL | For 35947 crucible | Each |
| 15401427 | 25 mL | For 35972 crucible | Each |
| 15441427 | 25 mL | For 35926 crucible | Each |
| 15481427 | 15 mL | For 35930 crucible | Each |
| 15481437 | 20 mL | For 35951 crucible | Each |
Product Code. 15401447
Brand: Thermo Scientific Chemicals 036111.KT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Description | For 36014 crucible |
| Material | Zirconium |
| Capacity (Metric) | 45 mL |
| Height (Metric) | Depth:29mm |
| Product Type | Zirconium Crucible Cover |