Learn More
Zirconium for AAS 1g/L HCl/HF matrix, Fisher chemical™
Catalog Number 12923305
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
7440-67-7
Zr
91.22
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
CHEBI:33342
zirconium
[Zr]
7440-67-7
Zr
91.22
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
CHEBI:33342
zirconium
[Zr]
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 1g/L ±0.002g/L
Packaging Bottle
Quantity 100 mL
Type Flame Single-element Standard Solution
Solvent or Matrix 5% HCl/0.05% HF
Physical Form Liquid
Chemical Name or Material Zirconium
compliance-icons
Product Identifier
  • Single-element Standard Solution for AAS. Zirconium (Zr) 1000mg/l in HCl 5%,HF 0.5%
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH210-Safety data sheet available on request.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.