missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIP8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20459-1-AP-20UL
This item is not returnable.
View return policy
Description
S39A8 encodes a member of the SLC39 family of solute-carrier genes, which show structural characteristics of zinc transporters. The encoded protein is glycosylated and found in the plasma membrane and mitochondria, and functions in the cellular import of zinc at the onset of inflammation. It is also thought to be the primary transporter of the toxic cation cadmium, which is found in cigarette smoke.Specifications
| ZIP8 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A8 | |
| BIGM103, PP3105, SLC39A8, ZIP 8, ZIP8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64116, 67547 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91W10, Q9C0K1 | |
| SLC39A8 | |
| ZIP8 Fusion Protein Ag14292 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction