Learn More
Emma Safety Footwear Zion XD Safety Shoes
Product Code. 15988301
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. 15988301
Brand: Emma Safety Footwear MM30186844
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical resistant with highly visible 3M™ reflective strip. The impenetrable sole protects feet from sharp and metal objects while the steel toe cap and reinforced polyurethane toe cover offer protection from falling objects. Microfiber components stand up to cement and remains flexible.
  • S3-Safety shoe with sole plate for wet work
  • Microfiber construction
  • GripForce™ Duo Heat Resistant Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Design Zion XD
Material Microfiber
Size 44