Learn More
Zinc Acetate Dihydrate, Supelco™
Product Code. 15845180
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15845180 1 kg 1kg
15835180 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15845180 Supplier Supelco Supplier No. 1088021000
 Restricted Item
Estimated Shipment: 26-11-2026
For analysis EMSURE™ ACS
Zinc acetate dihydrate ((CH3COO)2Zn · 2H2O) is hydrated form of zinc acetate. It is bidentate and has covalent character. It is commonly used as a zinc precursor in preparation of zinc oxide nanopowder.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5970-45-6
Specifications
Melting Point 237°C
Quantity 1 kg
Molecular Formula (CH3COO)2Zn · 2 H2O
MDL Number MFCD00066961
Synonym Zinc acetate dihydrate, Zinc acetate dihydrate
Molecular Weight (g/mol) 183.48 (anhydrous basis)
Grade Research
Density 1.74 g/cm3 at 20°C