missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZHX3 Polyclonal antibody apecifically detetects ZHX3 antigen in Human Samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | ZHX3 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 1100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
| Gene Accession No. | NM_015035 |
| Gene Symbols | ZHX3 |
| Host Species | Rabbit |
| Immunogen | ZHX3 fusion protein Ag30023 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?