missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29397-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
ZHX3 Polyclonal antibody apecifically detetects ZHX3 antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
ZHX3 | |
Polyclonal | |
Unconjugated | |
NM_015035 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human | |
Liquid |
Western Blot, ELISA | |
1100 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
ZHX3 | |
ZHX3 fusion protein Ag30023 | |
20 μL | |
23051 | |
Store at -20°C. Stable for one year after shipment. | |
IgG |