missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13118-2-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes an endosomal protein that belongs to the FYVE zinc finger family of proteins. The encoded protein is thought to regulate membrane trafficking in the endosome. This protein functions as a scaffold protein in the transforming growth factor-beta signaling pathway and is involved in positive and negative feedback regulation of the bone morphogenetic protein signaling pathway. Alternate splicing results in multiple transcript variants.Specifications
ZFYVE16 | |
Polyclonal | |
Unconjugated | |
ZFYVE16 | |
ENDOFIN, KIAA0305, ZFYVE16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
218441, 499508, 9765 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7Z3T8, Q80U44 | |
Zfyve16 | |
ZFYVE16 Fusion Protein Ag3811 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |