missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZFP90 may function as a repressor or silencer protein, and most likely exerts its repressing activity upon zinc-dependent binding to DNA. It may be involved in proper spermatogenesis by repressing the expression of genes unnecessary or incompatible with the maintece of a haploid cell state.
Specifications
Specifications
| Antigen | ZFP90 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.44 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | ZFP90 |
| Gene Accession No. | Q4V8A8, Q61967, Q8TF47 |
| Gene Alias | KIAA1954, NK10, Zfp 90, ZFP90, Zinc finger protein 90 homolog, ZNF756 |
| Gene Symbols | Zfp90 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?