missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP90 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26120-1-AP-150UL
This item is not returnable.
View return policy
Description
ZFP90 may function as a repressor or silencer protein, and most likely exerts its repressing activity upon zinc-dependent binding to DNA. It may be involved in proper spermatogenesis by repressing the expression of genes unnecessary or incompatible with the maintece of a haploid cell state.Specifications
| ZFP90 | |
| Polyclonal | |
| Unconjugated | |
| ZFP90 | |
| KIAA1954, NK10, Zfp 90, ZFP90, Zinc finger protein 90 homolog, ZNF756 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146198, 22751, 498945 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4V8A8, Q61967, Q8TF47 | |
| Zfp90 | |
| ZFP90 Fusion Protein Ag23381 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction