missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZFAT Polyclonal Antibody
GREENER_CHOICE

Product Code. 15742231 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15742231 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15742231 Supplier Invitrogen™ Supplier No. PA554297

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EPGGDIQQEA LGDQLQLVEE EFALQGVNAL KEEACPGDTQ LEEGRKEPEA PGEMPAPAVH LASPQAESTA LPPCELETT Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 61%.

This gene encodes a protein that likely binds DNA and functions as a transcriptional regulator involved in apoptosis and cell survival. This gene resides in a susceptibility locus for autoimmune thyroid disease (AITD) on chromosome 8q24. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFAT
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFAT
Gene Accession No. Q9P243
Gene Alias AITD3; Gm922; KIAA1485; ZFAT; ZFAT zinc finger 1; ZFAT1; Zfp406; zinc finger and AT hook domain containing; zinc finger and AT-hook domain containing; Zinc finger gene in AITD susceptibility region; zinc finger gene in autoimmune thyroid disease; zinc finger protein 403; zinc finger protein 406; Zinc finger protein ZFAT; zinc-finger gene in AITD susceptibility region; ZNF406
Gene Symbols ZFAT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFAT. Recombinant protein control fragment (Product #RP-93573).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57623
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.