Learn More
ZVG™ zetRoll™ Hand Towel Rolls
Description
The high-quality zetRoll™ disposable paper towel rolls are ideal for gastronomy, schools, care facilities, kitchens, and essential at gas stations for thorough removal of dirt, oil, and fuel residues. They provide a quick and convenient way to dry hands or quickly wipe surfaces or objects in the vicinity. All 2-ply and 3-ply paper towel rolls have point-to-point embossing.
- White
- High absorbency
- Hygienic hand drying
- With point-to-point embossing
Point-to-point embossing: In a novel manufacturing process, the individual layers of the paper towel are bonded together at multiple points across the entire surface. This creates a unified structure that is highly tear-resistant. This process combines the advantages of a thick wipe (stability) with those of a multi-layered towel (high absorbency).
Specifications
Specifications
| Product Type | Towel |
| Length (English) | 492.1 ft. |
| Length (Metric) | 150 m |
| Width (English) | 7.9 in. |
| Width (Metric) | 20 cm |
| Material | Cellulose |
| Color | White |
| Disposable | Single-use |
| Dimensions (L x W) | 150 m x 20 cm |
| Ply | 2-ply |