Learn More
3M™ Zeta Plus™ Activated Carbon Series Filter Cartridge
3M™ Zeta Plus™ Activated Carbon Series Filter Cartridge is a depth filter with carbon for removing impurities, color and particulates.
Brand: 3M™ 7100016149
Additional Details : Weight : 0.08000kg
Description
- Cartridges contain activated carbon immobilised within the filter matrix and eliminate need for handling bulk powdered carbon.
- Offered in a range of commercial activated carbons to match specific application requirements.
- Different media porosities match changing fluid viscosities and particle loading.
- Cartridges can be paired with Zeta Plus™ SP Series filter cartridges or capsules to optimise filtration and purification performance over a broad range of fluid challenges.
- Full range of scalable filters enables predictable performance at every stage of drug development from early stage discovery to commercial production.
- BC25 Laboratory capsules, media discs and stainless housings are ideal for selecting the best media grade using low challenge fluid volume.
Specifications
Filter Cartridge | |
C16MER53SP |