Learn More
3M™ Zeta Plus™ Activated Carbon Series Filter Cartridge
3M™ Zeta Plus™ Activated Carbon Series Filter Cartridge is a depth filter with carbon for removing impurities, color and particulates.
Brand:  3M™ 7100016149
Additional Details : Weight : 0.08000kg
Product Code. 17262474
  • 9900.00 NOK / Each
Estimated Shipment: 09-07-2024
Description
  • Cartridges contain activated carbon immobilised within the filter matrix and eliminate need for handling bulk powdered carbon.
  • Offered in a range of commercial activated carbons to match specific application requirements.
  • Different media porosities match changing fluid viscosities and particle loading.
  • Cartridges can be paired with Zeta Plus™ SP Series filter cartridges or capsules to optimise filtration and purification performance over a broad range of fluid challenges.
  • Full range of scalable filters enables predictable performance at every stage of drug development from early stage discovery to commercial production.
  • BC25 Laboratory capsules, media discs and stainless housings are ideal for selecting the best media grade using low challenge fluid volume.
Specifications
Specifications
Filter Cartridge
C16MER53SP