missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Zero-voltage Alarm Output for Growth Chambers

This Zero-voltage Alarm Output is designed for use with BINDER™ Growth Chambers. Audible signal can be turned on and off.

Brand:  BINDER™ 8012-1757

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 16444429

  • 19445.76 NOK / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket

Description

Description

  • For temperature (±2°C) and humidity (±5% RH)
  • Accessible via 6-pin DIN socket (max. 24 V, 2.5 A)
  • Audible signal can be switched off
Specifications

Specifications

Audible with ON/OFF Function
BINDER Chambers KBW 240/400 Growth Chambers with Light
6-pin DIN Socket (Max. 24 V, 2.5 A)
Alarm output, zero-voltage - for temperature (±2°C) and humidity (±5 % RH), accessible via 6-pin DIN socket (max. 24 V - 2.5 A), with audible signal that can be switched off
±5% RH
±2°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers