Learn More
Phenomenex™ Zebron™ Multi-Residue GC Column Kit, 0.32 mm ID, Includes: 2 Diff. Columns, Guard Column with γ-Splitter
Product Code. 31747368
Change view
Click to view available options
Length (Metric):
2 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
31747368 2 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31747368 Supplier Phenomenex™ Supplier No. KG08238
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
GC columns that deliver low bleed, excellent retention, and long column life for precise, reproducible results.
  • Guard Column
TRUSTED_SUSTAINABILITY
Specifications
Particle Size None
Length (Metric) 2 m
Packing Material Fused Silica
USP Type None
Stationary Phase Proprietary
Type GC Column
Diameter (Metric) Inner 0.32 mm