missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC13 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24759-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Palmitoyltransferase for HD and GAD2 (By similarity). Mediates Mg(2+) transport.Specifications
ZDHHC13 | |
Polyclonal | |
Unconjugated | |
ZDHHC13 | |
DHHC 13, HIP14 related protein, HIP14L, HIP3RP, Palmitoyltransferase ZDHHC13, ZDHHC13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
243983, 54503 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IUH4, Q9CWU2 | |
ZDHHC13 | |
ZDHHC13 Fusion Protein Ag21607 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |