Learn More
ZCL278, MedChemExpress
Product Code. 16455938
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16455938
Brand: Medchem Express HY1396310mg
MedChemExpress ZCL278 is a selective Cdc42 modulator that directly binds to Cdc42 and inhibits its functions with Kd of 11.4 μM for Cdc42-ZCL278 affinity in surface plasmon resonance (SPR) experiment.
ZCL278 is a selective Cdc42 modulator that directly binds to Cdc42 and inhibits its functions with Kd of 11.4 μM for Cdc42-ZCL278 affinity in surface plasmon resonance (SPR) experiment.
Specifications
Chemical Name or Material ZCL278
CAS 587841-73-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H19BrClN5O4S2
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (85.49 mM; Need ultrasonic)
SMILES O=C(NC(NC1=CC=C(S(=O)(NC2=NC(C)=CC(C)=N2)=O)C=C1)=S)COC3=CC=C(Br)C=C3Cl
Molecular Weight (g/mol) 584.89
Formula Weight 584.89
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.