missing translation for 'onlineSavingsMsg'
Learn More

ZBTB8OS Rabbit Polyclonal Antibody, Proteintech

Product Code. 30236102 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
30236102 150 μL 150µL
30233286 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 30236102

Brand: Proteintech 286081AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The ZBTB8OS antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZBTB8OS. This antibody recognizes human, mouse, rat antigen. The ZBTB8OS antibody has been validated for the following applications: WB, IHC, ELISA analysis.

Specifications

Antigen ZBTB8OS
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols ARCH; ARCH2; MGC62007
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339487
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 19 kDa
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.