Learn More
ZBTB2 Rabbit, anti-Human, Polyclonal, Proteintech
Product Code. 18019675 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
18019675 20 μL 20µL
18064505 150 μL 150µL
2 options
Product Code. 18019675
Brand: Proteintech 264361AP20UL
Rabbit Polyclonal Antibody
The ZBTB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZBTB2. This antibody recognizes human antigen. The ZBTB2 antibody has been validated for the following applications: IP, ELISA analysis.
Specifications
Antigen ZBTB2
Applications Immunoprecipitation, ELISA
Concentration 350 μg/mL
Gene Symbols KIAA1483, ZBTB2, ZNF437
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 57621
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG