missing translation for 'onlineSavingsMsg'
Learn More

ZBTB10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22944-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892135

  • 4593.76 NOK / 150µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a nuclear-localized protein that may be induced by p53 and regulates the cell cycle by inhibiting G1 to S phase progression. The encoded protein may interact with other cell cycle regulators.
Specifications

Specifications

ZBTB10
Polyclonal
Unconjugated
ZBTB10
RINZF, RINZFC, ZBTB10, Zinc finger protein RIN ZF
Rabbit
Antigen Affinity Chromatography
RUO
229055, 65986
-20°C
Liquid
Immunoprecipitation, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96DT7
ZBTB10
ZBTB10 Fusion Protein Ag19094
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers