Learn More
Labcon™ ZAP™ 1200 μL Aerosol Filter Pipette Tips for Sartorius™ Pipettes
These 1250 μL aerosol filter pipette tips have UltraFine™ points and come in racks.
Brand: Labcon™ 1194-965-006-9
Additional Details : Weight : 1.56200kg
Description
- Fits Sartorius™ 1200 μL Pipettes
- Protease, Human DNA, ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
- Validated Sterilization (ISO 11137) for use in Research and Medical labs
- Made in USA
Specifications
1200 μL | |
Sterile | |
Purple | |
88.9 mm | |
Protease, Human DNA, ATP, RNase/DNase, Endotoxin-free | |
E-beam | |
Rectangle | |
Single-use | |
No |
Filtered | |
Clear | |
Autoclavable | |
ISO 9001:2015, FDA CFR21 GMP, USP Class VI | |
Micropipetting | |
Rack | |
96 Tips/Rack, 6 Racks/Pk., 8 Pk./Cs. | |
Sartorius | |
Polypropylene Resin |