missing translation for 'onlineSavingsMsg'
Learn More

Zanubrutinib, MedChemExpress

Product Code. p-200097553
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16486107 5 mg 5mg
16423388 10 mg 10mg
16409916 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 16486107

Brand: Medchem Express HY101474A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zanubrutinib (BGB-3111) is a selective Bruton tyrosine kinase (Btk) inhibitor.

Zanubrutinib (BGB-3111) is a selective Bruton tyrosine kinase (Btk) inhibitor.

Specifications

Chemical Name or Material Zanubrutinib
CAS 1691249-45-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H29N5O3
Quantity 5 mg
Synonym BGB-3111
Solubility Information DMSO : 200 mg/mL (424.13 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble)
SMILES NC(C1=C2N(N=C1C3=CC=C(OC4=CC=CC=C4)C=C3)[C@H](C5CCN(C(C=C)=O)CC5)CCN2)=O
Molecular Weight (g/mol) 471.55
Formula Weight 471.55
Percent Purity 98.33%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.