Learn More
Zanubrutinib, MedChemExpress
Product Code. 16486107
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16486107
Brand: Medchem Express HY101474A
MedChemExpress Zanubrutinib (BGB-3111) is a selective Bruton tyrosine kinase (Btk) inhibitor.
Zanubrutinib (BGB-3111) is a selective Bruton tyrosine kinase (Btk) inhibitor.
Specifications
Chemical Name or Material Zanubrutinib
CAS 1691249-45-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H29N5O3
Quantity 5 mg
Synonym BGB-3111
Solubility Information DMSO : 200 mg/mL (424.13 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble)
SMILES NC(C1=C2N(N=C1C3=CC=C(OC4=CC=CC=C4)C=C3)[C@H](C5CCN(C(C=C)=O)CC5)CCN2)=O
Molecular Weight (g/mol) 471.55
Formula Weight 471.55
Percent Purity 98.33%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.