Learn More
Emma Safety Footwear Zandvoort XD Safety Shoes
Product Code. 15918751
Change view
Click to view available options
Size:
39
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15918751 39 1 pair
15928751 44 1 pair
15938751 45 1 pair
15948751 46 1 pair
15958751 47 1 pair
15968751 48 1 pair
6 options
Product Code. 15918751
Brand: Emma Safety Footwear MM54886639
Only null left
Add to basket
Add to basket
A mid-level boot with water repellant oiled Nubuck uppers, internal steel toe protection and an exceptional sticky grip even on wet and greasy surfaces. Primarily designed for Metal workers, the Zandvoort is used in outdoor occupations where a strong, safe, comfortable shoe is essential.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Design Zandvoort XD
Material Oiled Nubuck
Size 39