Learn More
Emma Safety Footwear Zandvoort Safety Shoes
Product Code. 15988691
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15988691 46 1 pair
15918691 37 1 pair
15928691 38 1 pair
15938691 39 1 pair
15948691 40 1 pair
15958691 41 1 pair
15968691 42 1 pair
15978691 43 1 pair
15998691 47 1 pair
15908701 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15988691 Supplier Emma Safety Footwear Supplier No. MM53884646
A mid-level boot with water repellant oiled Nubuck uppers, internal steel toe protection and an exceptional sticky grip even on wet and greasy surfaces. Primarily designed for Metal workers, the Zandvoort is used in outdoor occupations where a strong, safe, comfortable shoe is essential.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Brown
Design Zandvoort
Liner Material Full grain Nappa leather / Split leather
Material Oiled Nubuck
Size 46