Learn More
LNI Swissgas ZA TOTAL 15000
Shop All LNI Swissgas Products
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Description
- Zero air generator 15,000 cc/min HC/carbon dioxide/moisture removal.
- Needs source of dry and oil free air.
Specifications
Specifications
| Voltage | 100/115/220/230 V |
| Type | Zero Air Gas Generator |