Learn More
R&D Systems™ Z-VAD-FMK
Description
Z-VAD-FMK is a cell-permeable, irreversible pan-caspase inhibitor. Inhibits caspase processing and apoptosis induction in tumor cells in vitro (IC50 = 0.0015 - 5.8 mM). Active in vivo.
Specifications
Specifications
| Product Line | Tocris |
| Purity | >99% |
| CAS | 634911-81-9 |
| Description | Z-VAD-FMK |
| Molecular Weight (g/mol) | 467.49 |
| Product Type | Z-VAD-FMK |
| Quantity | 1 mg |
| Sequence | Z-Val-Ala-Asp(OMe)-fluoromethylketone |
For Research Use Only