Learn More
GVS Z-link respirator Zytec FR Chin Seal for PX5™ PAPR
Artikelnummer. 18419655
Skift visning
Klik for at se tilgængelige muligheder
Color:
Green
Pakningsstørrelse:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Color unitSize
18419655 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18419655 Leverandør GVS Leverandørnr. 1601031CE
  • ONLINE EXCLUSIVE PRICE
    7150.00 NOK
    6749.01 NOK / Pack of 1
    Save 400.99 NOK
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
The RPB Z-Link is the most versatile multi-purpose respirator on the market. Designed to be resilient in harsh industrial applications while practical to meet the demands of healthcare and life science settings.
The RPB Z-Link is the most versatile multi-purpose respirator on the market. Designed to be resilient in harsh industrial applications while practical to meet the demands of healthcare and life science settings. The Z-Link has been designed with your comfort and functionality in mind, ensuring no matter where you are working, you will be protected for the best moments.
  • Lightweight (880 g) and very comfortable using multi-adjustment helmet system.
  • Superior Comfort: the fully adjustable Comfort-Link fit system allows for total control of your sizing through supportive padding and the adjustable 360 degree head band and ratchet system.
  • Certified to EN 12941 1998 +A2:2008 (TH2 with PX5™ PAPR) and EN 166 2001 1 B T 9 3.
  • Technical Specification EN 397:2012 +A2:2012 (Impact Clauses 5.1.1, 5.1.2, 5.1.3) and
  • Optional Clauses: Very low temperature -300C, molten metal, lateral deformity, electrical properties 440v a.c.
  • Includes: 16-810 Safety Lens, 16-731 Chin Seal Zytec FR, 04-831 PX5™ Breathing Tube
  • To use with PAPR PX5™
Tekniske data
Color Green
Disposable Reusable
Material Polycarbonate
Product Type Respirator Helmet
Size One Size
With Exhalation Valve No
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.