Learn More
(Z)-Fluvoxamine, TRC
Product Code. 30135583
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135583 25 mg 25mg
30008694 1 mg 1mg
30006459 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135583 Supplier Toronto Research Chemicals Supplier No. TRCF60350525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89035-92-7
Molecular Formula C15 H21 F3 N2 O2
Molecular Weight (g/mol) 318.33
Synonym 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (1Z)-, 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (Z)-, 2-[[[(1Z)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethanamine, (Z)-Fluvoxamine
IUPAC Name 2-[(Z)-[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxyethanamine
SMILES COCCCC\C(=N\OCCN)\c1ccc(cc1)C(F)(F)F
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C15H21F3N2O2/c1-21-10-3-2-4-14(20-22-11-9-19)12-5-7-13(8-6-12)15(16,17)18/h5-8H,2-4,9-11,19H2,1H3/b20-14-
Formula Weight 318.1555
Percent Purity >95%
Chemical Name or Material (Z)-Fluvoxamine
Grade HPLC