missing translation for 'onlineSavingsMsg'
Learn More

(Z)-2-Decenoic acid, MedChemExpress

Product Code. 16435558
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
16435558 10 mg 10mg
16488257 5 mg 5mg
2 options
This item is not returnable. View return policy

Product Code. 16435558

Brand: Medchem Express HY13212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (Z)-2-decenoic acid (cis-2-Decenoic acid) is an unsaturated fatty acid produced by Pseudomonas aeruginosa. (Z)-2-decenoic acid induces a dispersion response in biofilms formed by a range of gram-negative bacteria, including P. aeruginosa, and by gram-positive bacteria. (Z)-2-decenoic acid inhibits biofilm development.

(Z)-2-decenoic acid (cis-2-Decenoic acid) is an unsaturated fatty acid produced by Pseudomonas aeruginosa. (Z)-2-decenoic acid induces a dispersion response in biofilms formed by a range of gram-negative bacteria, including P. aeruginosa, and by gram-positive bacteria. (Z)-2-decenoic acid inhibits biofilm development.

Specifications

Chemical Name or Material (Z)-2-Decenoic acid
CAS 15790-91-7
Color Colorless
Purity Grade Notes Research
Molecular Formula C10H18O2
Quantity 10 mg
Synonym cis-2-Decenoic acid
Solubility Information DMSO : 250 mg/mL (1468.43 mM; Need ultrasonic)
SMILES CCCCCCC/C=C\C(O)=O
Molecular Weight (g/mol) 170.25
Formula Weight 170.25
Percent Purity 97.0%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.