missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ YWHAB Recombinant Protein

Human YWHAB full-length ORF (AAH01359, 1 a.a. - 246 a.a.) recombinant protein with GST-tag at N-terminal

Brand:  Abnova™ H00007529-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141922

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein belonging to the 14-3-3 family of proteins, members of which mediate signal transduction by binding to phosphoserine-containing proteins.

  • Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide
  • Molecular weight: 52.8kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8 in the elution buffer
  • Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue

Sequence:
MTMDKSELVQKAKLAEQAERYDDMAAAMKAVTEQGHELSNEERNLLSVAYKNVVGARRSSWRVISSIEQKTERNEERQQMGKEYREKIEAELQDICNDVLELLDKYLIPNATQPESKVFYLKMKGDYFRYLSEVASGDNKQTTVSNSQQAYQEAFEISKKEMQPTHPIRLGLALNFSVFYYEILNSPEKACSLAKTAFDEAIAELDTLNEESYKDSTLIMQLLRDNLTLWTSENQGDEGDAGEGEN

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH01359
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
52.8
8
Glutathione Sepharose 4 Fast Flow
10 ug
MTMDKSELVQKAKLAEQAERYDDMAAAMKAVTEQGHELSNEERNLLSVAYKNVVGARRSSWRVISSIEQKTERNEERQQMGKEYREKIEAELQDICNDVLELLDKYLIPNATQPESKVFYLKMKGDYFRYLSEVASGDNKQTTVSNSQQAYQEAFEISKKEMQPTHPIRLGLALNFSVFYYEILNSPEKACSLAKTAFDEAIAELDTLNEESYKDSTLIMQLLRDNLTLWTSENQGDEGDAGEGEN
GW128/HS1/KCIP-1
YWHAB
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
7529
YWHAB (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
YWHAB
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers