missing translation for 'onlineSavingsMsg'
Learn More

YU238259, MedChemExpress

Product Code. 16460458
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16460458 5 mg 5mg
16493917 50 mg 50mg
16437776 100 mg 100mg
16427998 10 mg 10mg
16413726 25 mg 25mg
5 options
This item is not returnable. View return policy

Product Code. 16460458

Brand: Medchem Express HY19977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YU238259 is an inhibitor of homology-dependent DNA repair (HDR), used for cancer research.

YU238259 is an inhibitor of homology-dependent DNA repair (HDR), used for cancer research.

Specifications

Chemical Name or Material YU238259
CAS 1943733-16-1
Color White
Purity Grade Notes Research
Molecular Formula C22H22ClN3O4S
Quantity 5 mg
Solubility Information DMSO : 110 mg/mL (239.16 mM; Need ultrasonic)
SMILES COC1=CC=C(S(NCC2=CC=C(C(NCCC3=CC=C(Cl)C=N3)=O)C=C2)(=O)=O)C=C1
Molecular Weight (g/mol) 459.95
Formula Weight 459.95
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.