Learn More
Thermo Scientific Chemicals Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
Product Code. 15463825
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
50 mL
100 mL
200 mL
Diameter (Metric) Outer:
20mm
24mm
37mm
45mm
55mm
Height (Metric):
37mm
43mm
72mm
90mm
112mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer Height (Metric) unitSize
15463825 10 mL 24mm 43mm Each
15453825 5 mL 20mm 37mm Each
15473825 50 mL 37mm 72mm Each
15493825 200 mL 55mm 112mm Each
15483825 100 mL 45mm 90mm Each
5 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15463825
Brand: Thermo Scientific Chemicals 041274.KF
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Material Zirconium Oxide
Quantity 1 Piece
Capacity (Metric) 10 mL
Diameter (Metric) Outer 24mm
Height (Metric) 43mm
Product Type Yttria Stabilized Zirconium Oxide Cylindrical Crucibles