missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™

Catalog Number 12997443
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mL
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
7440-64-4
Yb
173.05
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
23992
CHEBI:33381
ytterbium
[Yb]
This item is not returnable. View return policy
7440-64-4
Yb
173.05
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
23992
CHEBI:33381
ytterbium
[Yb]

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10mg/L
Packaging Bottle
Quantity 50 mL
Solvent or Matrix HNO3
Chemical Name or Material Ytterbium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'