missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YTHDF3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25537-1-AP-150UL
This item is not returnable.
View return policy
Description
Product No. 25537-1-AP specifically reacts with YTHDF3 protein.
This gene encodes a member of the YTH (YT521-B homology) domain protein family. The YTH domain is common in eukaryotes, is often found in the middle of the protein sequence, and may function in binding to RNA. Alternative splicing results in multiple transcript variants.Specifications
| YTHDF3 | |
| Polyclonal | |
| Unconjugated | |
| YTHDF3 | |
| YTH domain family protein 3, YTH domain family, member 3, YTHDF3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229096, 253943 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z739, Q8BYK6 | |
| YTHDF3 | |
| YTHDF3-specific Fusion Protein Ag22374 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction