missing translation for 'onlineSavingsMsg'
Learn More

YTHDF3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25537-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16844105

  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product No. 25537-1-AP specifically reacts with YTHDF3 protein.

This gene encodes a member of the YTH (YT521-B homology) domain protein family. The YTH domain is common in eukaryotes, is often found in the middle of the protein sequence, and may function in binding to RNA. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

YTHDF3
Polyclonal
Unconjugated
YTHDF3
YTH domain family protein 3, YTH domain family, member 3, YTHDF3
Rabbit
Antigen Affinity Chromatography
RUO
229096, 253943
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q7Z739, Q8BYK6
YTHDF3
YTHDF3-specific Fusion Protein Ag22374
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers