Learn More
MP Biomedicals™ YPD (YEPD) Broth ( Powder)
Product Code. 11337749 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
454 g
1000 g
Unit Size:
1kg
454g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337749 454 g 454g
11347749 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11337749 Supplier MP Biomedicals™ Supplier No. 114001022
  • ONLINE EXCLUSIVE PRICE
    1550.00 NOK
    1473.83 NOK / 454g
    Save 76.17 NOK
    4% Off
 Restricted Item
In Stock
Rich medium for the growth of S. cerevisiae in liquid medium
  • Contains, dextrose, peptone, and yeast extract
  • pH 6.5 at 300°C
  • Bulk powder packaging
Formulation
  • 50g/L
  • Contents/liter: 20g peptone, 10g yeast extract, 20g dextrose
Yeast Cell Culture
Specifications
Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Microbiology Media
Formulation Casein Peptone type-M 20g/L, Yeast extract 10g/L, Dextrose 20g/L
For Use With (Application) For growth of Saccharomyces cerevisiae in liquid medium
Packaging Type Bulk Powder
Quantity 454 g