Learn More
YK11, MedChemExpress
Product Code. 16474138
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16474138 10 mg 10mg
16465106 100 mg 100mg
16410497 50 mg 50mg
3 options
Product Code. 16474138
Brand: Medchem Express HY10748010mg
MedChemExpress YK11 is a partial agonist of androgen receptor, with osteogenic activity.
YK11 is a partial agonist of androgen receptor, with osteogenic activity.
Specifications
Chemical Name or Material YK11
CAS 1370003-76-1
Color White
Purity Grade Notes Research
Molecular Formula C25H34O6
Quantity 10 mg
Solubility Information DMSO : ≥ 64 mg/mL (148.65 mM)
SMILES O=C1CCC2C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@@]4(OC(C)(OC)O/5)C5=C/C(OC)=O)=C1
Molecular Weight (g/mol) 430.53
Formula Weight 430.53
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism