missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ YIF1B Polyclonal Antibody
GREENER_CHOICE

Product Code. 17120824 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17120824 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17120824 Supplier Invitrogen™ Supplier No. PA5112961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

YIF1B Polyclonal Antibody for ICC/IF, IHC (P), ELISA

YIF1B belongs to the YIF1 family. It is a multi-pass membrane protein. The functions of YIF1B remain unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YIF1B
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 3.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene YIF1B
Gene Accession No. Q5BJH7
Gene Alias 9430029K10Rik; FinGER8; PP4519; Protein YIF1B; UNQ3073/PRO9905; YIF1B; Yip1 interacting factor homolog B; Yip1 interacting factor homolog B (S. cerevisiae); Yip1 interacting factor homolog B, membrane trafficking protein; Yip1b; YIP1-interacting factor homolog B
Gene Symbols YIF1B
Host Species Rabbit
Immunogen Recombinant Human Protein YIF1B protein (1-153AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90522
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.