missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yersinia Selective Agar Base (Dehydrated)

Product Code. p-200112465
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10482643

Brand: Thermo Scientific™ CM0653B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples with this selective medium

Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples with Thermo Scientific™ Oxoid™ Yersinia Selective Agar Base (Dehydrated) when used with Yersinia Selective Supplement (SR0109).

Yersinia enterocolitica is a bacterial species in the family Enterobacteriaceae that most often causes enterocolitis, acute diarrhea, terminal ileitis, mesenteric lymphadenitis, and pseudoappendicitis but, if it spreads systemically, can also result in fatal sepsis.

Yersinia Selective Agar Base and the selective supplement SR0109 have been developed specifically for the optimum growth and recovery of Yersinia enterocolitica after 18-24 hours incubation at 32°C.

The typical colonies of Yersinia enterocolitica will develop as a red bull’s-eye surrounded by a transparent border and will vary considerably among serotypes in colony size, smoothness and the ratio of the border to center diameter. Most other organisms that are capable of growing will produce larger colonies (>2mm in diameter) with diffuse pinkish centers and opaque outer zones. Serratia liquefaciens, Citrobacter freundii and Enterobacter agglomerans may give a colonial morphology resembling Yersinia enterocolitica. These organisms can be differentiated from Yersinia enterocolitica by biochemical tests.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Yersinia Selective Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 8.6L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.